missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6618906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine | |
C22H25N3O5 | |
fmoc-hocit-oh, fmoc-l-homocitrulline, fmoc-hcit-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-ureidohexanoic acid, fmoc-homocit-oh, 2s-6-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1383, fmoc-lys carbamoyl-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homocitrulline, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-6-ureidohexanoic acid | |
NC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
411.46 | |
411.46 | |
5 g |
201485-17-8 | |
MFCD00237387 | |
VJZUCXPETVPQIB-TWYLJJHKNA-N | |
(2S)-6-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid | |
7019813 | |
95% | |
Solid |
Chemical Identifiers
201485-17-8 | |
411.46 | |
VJZUCXPETVPQIB-TWYLJJHKNA-N | |
7019813 | |
NC(=O)NCCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
C22H25N3O5 | |
MFCD00237387 | |
fmoc-hocit-oh, fmoc-l-homocitrulline, fmoc-hcit-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-ureidohexanoic acid, fmoc-homocit-oh, 2s-6-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1383, fmoc-lys carbamoyl-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-homocitrulline, s-2-9h-fluoren-9-yl methoxy carbonyl-amino-6-ureidohexanoic acid | |
(2S)-6-(carbamoylamino)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)hexanoic acid |
Safety and Handling
TSCA : No
storageNote1 : -30° to -10°C
Recommended Storage : Store at -20°C
RUO – Research Use Only