Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
CAS 167393-62-6
Formula Weight 624.77
MDL Number MFCD00237166
Synonym Fmoc-Lys(Mtt)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6362903

  SDS
alfa aesar™
H6362903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6362906

  SDS
alfa aesar™
H6362906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 624.77
Chemical Name or Material Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-L-lysine
Physical Form Solid
Assay 95%
MDL Number MFCD00237166
CAS 167393-62-6
Synonym Fmoc-Lys(Mtt)-OH
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C