missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Fmoc-N^e-(4-methyltrityl)-D-lysine, 97%, Thermo Scientific Chemicals
$48.37 - $569.37
Chemical Identifiers
CAS | 198544-94-4 |
---|---|
Molecular Formula | C41H40N2O4 |
Molecular Weight (g/mol) | 624.78 |
MDL Number | MFCD26793593 |
InChI Key | YPTNAIDIXCOZAJ-KXQOOQHDSA-N |
Synonym | r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-4-methyltrityl-d-lysine, ambotzfaa1130, fmoc-dlys mtt-oh, fmoc-d-lys meotrt-oh, fmoc-d-lysine mtt-oh, fmoc-n-epsilon-methyltrityl-d-lysine, n-a-fmoc-n-d-methyltrityl-d-lysine, fmoc-n-epsilon-4-methoxy trityl-d-lysine, n-alpha-fmoc-n-epsilon-4-methyltrityl-d-lysine |
PubChem CID | 51340501 |
IUPAC Name | (2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid |
SMILES | CC1=CC=C(C=C1)C(NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH66803MD
|
Thermo Scientific Chemicals
H66803MD |
250 mg |
Each for $48.37
|
|
|||||
AAH6680303
|
Thermo Scientific Chemicals
H6680303 |
1 g |
Each for $134.29
|
|
|||||
AAH6680306
|
Thermo Scientific Chemicals
H6680306 |
5 g |
Each for $569.37
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
198544-94-4 | |
624.78 | |
YPTNAIDIXCOZAJ-KXQOOQHDSA-N | |
51340501 | |
CC1=CC=C(C=C1)C(NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 |
C41H40N2O4 | |
MFCD26793593 | |
r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-4-methyltrityl-d-lysine, ambotzfaa1130, fmoc-dlys mtt-oh, fmoc-d-lys meotrt-oh, fmoc-d-lysine mtt-oh, fmoc-n-epsilon-methyltrityl-d-lysine, n-a-fmoc-n-d-methyltrityl-d-lysine, fmoc-n-epsilon-4-methoxy trityl-d-lysine, n-alpha-fmoc-n-epsilon-4-methyltrityl-d-lysine | |
(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid |
Specifications
198544-94-4 | |
MFCD26793593 | |
YPTNAIDIXCOZAJ-KXQOOQHDSA-N | |
(2R)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-{[(4-methylphenyl)diphenylmethyl]amino}hexanoic acid | |
51340501 | |
98% | |
Solid |
C41H40N2O4 | |
r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-diphenyl p-tolyl methyl amino hexanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-4-methyltrityl-d-lysine, ambotzfaa1130, fmoc-dlys mtt-oh, fmoc-d-lys meotrt-oh, fmoc-d-lysine mtt-oh, fmoc-n-epsilon-methyltrityl-d-lysine, n-a-fmoc-n-d-methyltrityl-d-lysine, fmoc-n-epsilon-4-methoxy trityl-d-lysine, n-alpha-fmoc-n-epsilon-4-methyltrityl-d-lysine | |
CC1=CC=C(C=C1)C(NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O)(C1=CC=CC=C1)C1=CC=CC=C1 | |
624.78 | |
624.78 | |
250 mg | |
Nalpha-Fmoc-Nepsilon-(4-methyltrityl)-D-lysine |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only