Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-Ndelta-trityl-L-glutamine pentafluorophenyl ester, 97%

$320.70 - $320.70

Chemical Identifiers

Chemical Name or Material Nalpha-Fmoc-Ndelta-trityl-L-glutamine pentafluorophenyl ester
CAS 132388-65-9
Molecular Formula C45H33F5N2O5
Formula Weight 776.75g/mol
MDL Number MFCD00800874
Synonym Fmoc-Gln(Trt)-OPfp
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2718206

 
alfa aesar™
H27182-06
5g Each for $320.70
Description & Specifications

Specifications

Melting Point 183° to 184°C
Quantity 5g
Optical Rotation −16° (c=1 in THF)
Chemical Name or Material Nalpha-Fmoc-Ndelta-trityl-L-glutamine pentafluorophenyl ester
CAS 132388-65-9
Assay 97%
Molecular Formula C45H33F5N2O5
Formula Weight 776.75g/mol
MDL Number MFCD00800874
Synonym Fmoc-Gln(Trt)-OPfp
Recommended Storage 2° to 8°C