Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-L-histidine, 98%

Chemical Identifiers

Chemical Name or Material Nalpha-Fmoc-L-histidine
CAS 116611-64-4
Formula Weight 377.4
MDL Number MFCD00190885
Synonym Fmoc-His-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6640706

 
alfa aesar™
H6640706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6640714

 
alfa aesar™
H6640714
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 377.4
Chemical Name or Material Nalpha-Fmoc-L-histidine
MDL Number MFCD00190885
Synonym Fmoc-His-OH
CAS 116611-64-4
Assay 98%
Physical Form Solid
Storage Note 1 -30° to -10°C
Quantity 5g
Safety and Handling

-30° to -10°C