missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Fmoc-L-arginine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6666506
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nalpha-Fmoc-L-arginine | |
White | |
MFCD00051770 | |
fmoc-arg-oh, fmoc-l-arginine, nalpha-fmoc-l-arginine, alfa-fmoc-l-arginine, fmoc-l-arg-oh, n-a-fmoc-l-arginine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-guanidino-pentanoic acid, 2s-5-amidinoamino-2-fluoren-9-ylmethoxy carbonylamino pentanoic acid, 2s-5-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-arg | |
NC(=[NH2+])NCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O | |
396.45 | |
396.45 | |
5 g | |
Powder |
91000-69-0 | |
C21H24N4O4 | |
4828015 | |
DVBUCBXGDWWXNY-SFHVURJKSA-N | |
(2S)-5-{[amino(iminiumyl)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoate | |
2724631 | |
95% | |
Moisture sensitive |
Chemical Identifiers
91000-69-0 | |
396.45 | |
DVBUCBXGDWWXNY-SFHVURJKSA-N | |
2724631 | |
NC(=[NH2+])NCCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O |
C21H24N4O4 | |
MFCD00051770 | |
fmoc-arg-oh, fmoc-l-arginine, nalpha-fmoc-l-arginine, alfa-fmoc-l-arginine, fmoc-l-arg-oh, n-a-fmoc-l-arginine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-guanidino-pentanoic acid, 2s-5-amidinoamino-2-fluoren-9-ylmethoxy carbonylamino pentanoic acid, 2s-5-carbamimidamido-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-arg | |
(2S)-5-{[amino(iminiumyl)methyl]amino}-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)pentanoate |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only