Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Fmoc-3-(Fmoc-amino)-L-alanine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Fmoc-3-(Fmoc-amino)-L-alanine
CAS 201473-90-7
Formula Weight 548.6
MDL Number MFCD00237005
Synonym Fmoc-Dap(Fmoc)-OH; (S)-2,3-Bis(Fmoc-amino)propionic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6639803

 
alfa aesar™
H6639803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6639806

 
alfa aesar™
H6639806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66398MD

 
alfa aesar™
H66398MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 548.6
Chemical Name or Material Nalpha-Fmoc-3-(Fmoc-amino)-L-alanine
MDL Number MFCD00237005
CAS 201473-90-7
Physical Form Crystalline
Assay 95%
Color Off-white
Storage Note 1 2° to 8°C
Synonym Fmoc-Dap(Fmoc)-OH; (S)-2,3-Bis(Fmoc-amino)propionic acid
Quantity 1g
Safety and Handling

2° to 8°C