missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nalpha-Carbobenzoxy-DL-asparagine, TCI America™

$18.33 - $68.75

Chemical Identifiers

CAS 29880-22-6
Molecular Formula C12H14N2O5
Molecular Weight (g/mol) 266.253
MDL Number MFCD00065101
InChI Key FUCKRCGERFLLHP-UHFFFAOYSA-N
Synonym z-dl-asparagine, z-dl-asn-oh, n-carbobenzyloxy-d-asparagine, n-carbobenzoxy-dl-asparagine, 2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid, nalpha-carbobenzoxy-dl-asparagine, carbobenzoxy-l-asparagine, 4-amino-4-oxo-2-phenylmethoxycarbonylamino butanoic acid, asparagine, n 2-benzyl ester, l, benzyloxycarbonyl-asparagine
PubChem CID 246565
IUPAC Name 4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C06611G View Documents TCI America
C06611G
1g Each for $18.33
 
C066110G View Documents TCI America
C066110G
10g Each for $68.75
 
Specifications

Chemical Identifiers

29880-22-6
266.253
FUCKRCGERFLLHP-UHFFFAOYSA-N
246565
C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O
C12H14N2O5
MFCD00065101
z-dl-asparagine, z-dl-asn-oh, n-carbobenzyloxy-d-asparagine, n-carbobenzoxy-dl-asparagine, 2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid, nalpha-carbobenzoxy-dl-asparagine, carbobenzoxy-l-asparagine, 4-amino-4-oxo-2-phenylmethoxycarbonylamino butanoic acid, asparagine, n 2-benzyl ester, l, benzyloxycarbonyl-asparagine
4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid

Specifications

White
1g
C12H14N2O5
z-dl-asparagine, z-dl-asn-oh, n-carbobenzyloxy-d-asparagine, n-carbobenzoxy-dl-asparagine, 2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid, nalpha-carbobenzoxy-dl-asparagine, carbobenzoxy-l-asparagine, 4-amino-4-oxo-2-phenylmethoxycarbonylamino butanoic acid, asparagine, n 2-benzyl ester, l, benzyloxycarbonyl-asparagine
C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O
266.253
266.25
≥99.0% (T)
165°C
29880-22-6
MFCD00065101
FUCKRCGERFLLHP-UHFFFAOYSA-N
4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid
246565
Crystal-Powder at 20°C
Nalpha-Carbobenzoxy-DL-asparagine
Safety and Handling

Safety and Handling

TSCA : No