missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Carbobenzoxy-DL-asparagine 99.0+%, TCI America™
Supplier: TCI America C066110G
Specifications
Nalpha-Carbobenzoxy-DL-asparagine | |
165°C | |
C12H14N2O5 | |
10 g | |
FUCKRCGERFLLHP-UHFFFAOYSA-N | |
4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid | |
246565 | |
≥99.0% (T) |
29880-22-6 | |
White | |
MFCD00065101 | |
z-dl-asparagine, z-dl-asn-oh, n-carbobenzyloxy-d-asparagine, n-carbobenzoxy-dl-asparagine, 2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid, nalpha-carbobenzoxy-dl-asparagine, carbobenzoxy-l-asparagine, 4-amino-4-oxo-2-phenylmethoxycarbonylamino butanoic acid, asparagine, n 2-benzyl ester, l, benzyloxycarbonyl-asparagine | |
C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O | |
266.253 | |
266.25 | |
Crystalline Powder |
Chemical Identifiers
29880-22-6 | |
266.253 | |
FUCKRCGERFLLHP-UHFFFAOYSA-N | |
246565 | |
C1=CC=C(C=C1)COC(=O)NC(CC(=O)N)C(=O)O |
C12H14N2O5 | |
MFCD00065101 | |
z-dl-asparagine, z-dl-asn-oh, n-carbobenzyloxy-d-asparagine, n-carbobenzoxy-dl-asparagine, 2-benzyloxy carbonyl amino-3-carbamoylpropanoic acid, nalpha-carbobenzoxy-dl-asparagine, carbobenzoxy-l-asparagine, 4-amino-4-oxo-2-phenylmethoxycarbonylamino butanoic acid, asparagine, n 2-benzyl ester, l, benzyloxycarbonyl-asparagine | |
4-amino-4-oxo-2-(phenylmethoxycarbonylamino)butanoic acid |
Safety and Handling
TSCA : No