Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine
CAS 186698-61-3
Formula Weight 526.65
MDL Number MFCD00236822
Synonym Boc-D-Arg(Pbf)-OH; Nα-Boc-Nω-Pbf-D-arginine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6299703

 
alfa aesar™
H6299703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62997MD

 
alfa aesar™
H62997MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 526.65
Chemical Name or Material Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-D-arginine
Physical Form Solid
Assay 95%
MDL Number MFCD00236822
CAS 186698-61-3
Synonym Boc-D-Arg(Pbf)-OH; Nα-Boc-Nω-Pbf-D-arginine
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C