missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine, 96%, Alfa Aesar™

Manufacturer:  Alfa Aesar H6214806

 View more versions of this product

Catalog No. AAH6214806



Description

Description

Applications
Nalpha-Boc-Nµ-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine is used as a triflating reagent.

Solubility
Soluble in water or 1% acetic acid

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Nalpha-Boc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-sulfonyl)-L-arginine
200124-22-7
MFCD00236821
CVFXPOKENLGCID-KRWDZBQOSA-N
(2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
11341472
96%
5g
C24H38N4O7S
boc-arg pbf-oh, 2s-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2s-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid, s-2-tert-butoxycarbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, n-boc-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sufonyl-l-arginine, ambotzbaa1067, boc-arg pbf-0h, n,a-boc-n, o-pbf-l-arginine, boc-arg pbf-oh tlc, n,a-boc-n, o-2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl-l-arginine
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C
526.649
526.65

Chemical Identifiers

200124-22-7
526.649
CVFXPOKENLGCID-KRWDZBQOSA-N
11341472
CC1=C2C(=C(C(=C1C)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N)C)CC(O2)(C)C
C24H38N4O7S
MFCD00236821
boc-arg pbf-oh, 2s-2-tert-butoxycarbonyl amino-5-n'-2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-ylsulfonyl carbamimidamido pentanoic acid, 2s-5-e-amino 2,2,4,6,7-pentamethyl-3h-1-benzofuran-5-sulfonamido methylidene amino-2-tert-butoxycarbonyl amino pentanoic acid, s-2-tert-butoxycarbonyl amino-5-3-2,2,4,6,7-pentamethyl-2,3-dihydrobenzofuran-5-yl sulfonyl guanidino pentanoic acid, n-boc-n'-2,2,4,6,7-pentamethyldihydrobenzofuran-5-sufonyl-l-arginine, ambotzbaa1067, boc-arg pbf-0h, n,a-boc-n, o-pbf-l-arginine, boc-arg pbf-oh tlc, n,a-boc-n, o-2,2,4,6,7-pentamethyl-dihydrobenzofuran-5-sulfonyl-l-arginine
(2S)-5-[[amino-[(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-yl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents