missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Ngamma-xanthyl-L-asparagine, 98%

$25.91 - $86.81

Chemical Identifiers

CAS 65420-40-8
Molecular Formula C22H24N2O6
Molecular Weight (g/mol) 412.442
MDL Number MFCD00066157
InChI Key YMGDQLXBNMRJMR-HNNXBMFYSA-N
Synonym boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
PubChem CID 7017974
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
SMILES CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6369006 View Documents Alfa Aesar™
H6369006
5g Each for $25.91
 
AAH6369014 View Documents Alfa Aesar™
H6369014
25g Each for $86.81
 
Specifications

Chemical Identifiers

65420-40-8
412.442
YMGDQLXBNMRJMR-HNNXBMFYSA-N
7017974
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
C22H24N2O6
MFCD00066157
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid

Specifications

65420-40-8
MFCD00066157
boc-asn xan-oh, s-4-9h-xanthen-9-yl amino-2-tert-butoxycarbonyl amino-4-oxobutanoic acid, n-alpha-t-boc-n-beta-xanthyl-l-asparagine, n-boc-n-xanthyl-l-asparagine, 2s-2-tert-butoxycarbonyl amino-3-9h-xanthen-9-yl carbamoyl propanoic acid, ambotzbaa1070, pubchem18926, nalpha-boc-nbeta-xanthenyl-l-asparagine, nalpha-boc-nbeta-xanthenyl-l-asparagine, n alpha-boc-n gamma-9-xanthenyl-l-asparagine
CC(C)(C)OC(=O)NC(CC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
412.442
412.44
98%
177°C to 181°C (decomposition)
Nalpha-Boc-Ngamma-xanthyl-L-asparagine
C22H24N2O6
5172403
YMGDQLXBNMRJMR-HNNXBMFYSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxo-4-(9H-xanthen-9-ylamino)butanoic acid
7017974
Solid
Orange
5g
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C