Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine
CAS 16965-06-3
Formula Weight 342.31
MDL Number MFCD00037104
Synonym Boc-Lys(Tfa)-OH; (S)-2-(Boc-amino)-6-(2,2,2-trifluoroacetamido)hexanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6314303

 
alfa aesar™
H6314303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6314306

 
alfa aesar™
H6314306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6314314

 
alfa aesar™
H6314314
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 342.31
Assay 95%
MDL Number MFCD00037104
CAS 16965-06-3
Quantity 1g
Storage Note 1 -30° to -10°C
Chemical Name or Material Nalpha-Boc-Nepsilon-trifluoroacetyl-L-lysine
Physical Form Powder
Synonym Boc-Lys(Tfa)-OH; (S)-2-(Boc-amino)-6-(2,2,2-trifluoroacetamido)hexanoic acid
Color White

Applications
N alpha-Boc-N epsilon-trifluoroacetyl-L-lysine, is used as a fine chemical intermediate.

Solubility
Soluble in DMSO.

Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Safety and Handling

-30° to -10°C