Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Nepsilon-trifluoroacetyl-D-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Boc-Nepsilon-trifluoroacetyl-D-lysine
CAS 96561-04-5
Formula Weight 342.32
MDL Number MFCD00236857
Synonym Boc-D-Lys(Tfa)-OH; Nα-Boc-Nε-(2,2,2-trifluoroacetyl)-D-lysine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6660303

 
alfa aesar™
H6660303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66603MD

 
alfa aesar™
H66603MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 342.32
Chemical Name or Material Nalpha-Boc-Nepsilon-trifluoroacetyl-D-lysine
Assay 95%
CAS 96561-04-5
Physical Form Solid
MDL Number MFCD00236857
Storage Note 1 -30° to -10°C
Synonym Boc-D-Lys(Tfa)-OH; Nα-Boc-Nε-(2,2,2-trifluoroacetyl)-D-lysine
Quantity 1g
Safety and Handling

-30° to -10°C