Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Nepsilon-formyl-L-lysine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Boc-Nepsilon-formyl-L-lysine
CAS 2483-47-8
Formula Weight 274.32
MDL Number MFCD00037361
Synonym Boc-Lys(For)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6663503

 
alfa aesar™
H6663503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6663506

 
alfa aesar™
H6663506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 274.32
Chemical Name or Material Nalpha-Boc-Nepsilon-formyl-L-lysine
Physical Form Powder
CAS 2483-47-8
Assay 95%
MDL Number MFCD00037361
Quantity 1g
Synonym Boc-Lys(For)-OH
Color Off-white to white
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C