Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Nepsilon-Fmoc-L-lysine methyl ester, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Boc-Nepsilon-Fmoc-L-lysine methyl ester
CAS 133628-28-1
Formula Weight 482.58
MDL Number MFCD00236856
Synonym Boc-Lys(Fmoc)-OMe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6671203

  SDS
alfa aesar™
H6671203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66712MD

  SDS
alfa aesar™
H66712MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 482.58
Chemical Name or Material Nalpha-Boc-Nepsilon-Fmoc-L-lysine methyl ester
MDL Number MFCD00236856
CAS 133628-28-1
Physical Form Powder
Assay 95%
Synonym Boc-Lys(Fmoc)-OMe
Storage Note 1 -30° to -10°C
Color White to off-white
Quantity 1g

Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Safety and Handling

-30° to -10°C