missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Nepsilon-benzyloxycarbonyl-D-lysine, 98%

$93.15 - $346.97

Chemical Identifiers

CAS 76477-42-4
Molecular Formula C19H28N2O6
Molecular Weight (g/mol) 380.441
MDL Number MFCD00038262
InChI Key BDHUTRNYBGWPBL-OAHLLOKOSA-N
Synonym 2r-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid
PubChem CID 7269335
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6209603 View Documents Alfa Aesar™
H6209603
1g Each for $93.15
 
AAH6209606 View Documents Alfa Aesar™
H6209606
5g Each for $346.97
 
Specifications

Chemical Identifiers

76477-42-4
380.441
BDHUTRNYBGWPBL-OAHLLOKOSA-N
7269335
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)O
C19H28N2O6
MFCD00038262
2r-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid

Specifications

76477-42-4
MFCD00038262
BDHUTRNYBGWPBL-OAHLLOKOSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid
7269335
Solid
1g
C19H28N2O6
2r-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)O
380.441
380.44
98%
Nalpha-Boc-Nepsilon-benzyloxycarbonyl-D-lysine