missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Nepsilon-benzyloxycarbonyl-D-lysine, 98%

Manufacturer:  Alfa Aesar™ H6209606

 View more versions of this product

Catalog No. AAH6209606



Specifications

Specifications

Nalpha-Boc-Nepsilon-benzyloxycarbonyl-D-lysine
C19H28N2O6
2r-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)O
380.441
380.44
98%
76477-42-4
MFCD00038262
BDHUTRNYBGWPBL-OAHLLOKOSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid
7269335
Solid
5g

Chemical Identifiers

76477-42-4
380.441
BDHUTRNYBGWPBL-OAHLLOKOSA-N
7269335
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1=CC=CC=C1)C(=O)O
C19H28N2O6
MFCD00038262
2r-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid
Documents