Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Nepsilon-(2-chlorobenzyloxycarbonyl)-D-lysine, 98%

$181.64 - $181.64

Chemical Identifiers

Chemical Name or Material Nalpha-Boc-Nepsilon-(2-chlorobenzyloxycarbonyl)-D-lysine
CAS 57096-11-4
Formula Weight 414.88
MDL Number MFCD00065585
Beilstein 5913148
Synonym Boc-D-Lys(2-Cl-Z)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6306906

  SDS
alfa aesar™
H6306906
5g Each for $181.64

AAH6306903

  SDS
alfa aesar™
H6306903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 414.88
Chemical Name or Material Nalpha-Boc-Nepsilon-(2-chlorobenzyloxycarbonyl)-D-lysine
Physical Form Powder
Assay 98%
MDL Number MFCD00065585
CAS 57096-11-4
Beilstein 5913148
Synonym Boc-D-Lys(2-Cl-Z)-OH
Color White
Quantity 5g