missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Ndelta-xanthyl-D-glutamine, 98%

$41.18 - $487.31

Chemical Identifiers

CAS 99092-88-3
Molecular Formula C23H26N2O6
Molecular Weight (g/mol) 426.469
MDL Number MFCD00151873
InChI Key HOIIDGIJEPOSLL-MRXNPFEDSA-N
Synonym boc-d-gln xan-oh, ambotzbaa1180, n-alpha-boc-n-delta-xanthyl-d-glutamine, n-alpha-t-butyloxycarbonyl-n-gamma-xantyl-d-glutamine, n~2~-tert-butoxycarbonyl-n-9h-xanthen-9-yl-d-glutamine, 2r-2-tert-butoxycarbonyl amino-4-9h-xanthen-9-yl carbamoyl butanoic acid
PubChem CID 40424385
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-(9H-xanthen-9-ylamino)pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66150MD View Documents Alfa Aesar™
H66150MD
250mg Each for $41.18
 
AAH6615003 View Documents Alfa Aesar™
H6615003
1g Each for $122.45
 
AAH6615006 View Documents Alfa Aesar™
H6615006
5g Each for $487.31
 
Specifications

Chemical Identifiers

99092-88-3
426.469
HOIIDGIJEPOSLL-MRXNPFEDSA-N
40424385
CC(C)(C)OC(=O)NC(CCC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
C23H26N2O6
MFCD00151873
boc-d-gln xan-oh, ambotzbaa1180, n-alpha-boc-n-delta-xanthyl-d-glutamine, n-alpha-t-butyloxycarbonyl-n-gamma-xantyl-d-glutamine, n~2~-tert-butoxycarbonyl-n-9h-xanthen-9-yl-d-glutamine, 2r-2-tert-butoxycarbonyl amino-4-9h-xanthen-9-yl carbamoyl butanoic acid
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-(9H-xanthen-9-ylamino)pentanoic acid

Specifications

99092-88-3
MFCD00151873
HOIIDGIJEPOSLL-MRXNPFEDSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-(9H-xanthen-9-ylamino)pentanoic acid
40424385
Powder
White
Nalpha-Boc-Ndelta-xanthyl-D-glutamine
C23H26N2O6
boc-d-gln xan-oh, ambotzbaa1180, n-alpha-boc-n-delta-xanthyl-d-glutamine, n-alpha-t-butyloxycarbonyl-n-gamma-xantyl-d-glutamine, n~2~-tert-butoxycarbonyl-n-9h-xanthen-9-yl-d-glutamine, 2r-2-tert-butoxycarbonyl amino-4-9h-xanthen-9-yl carbamoyl butanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
426.469
426.47
98%
250mg
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C