missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-Ndelta-Fmoc-L-ornithine, 95%

$28.84 - $300.72

Chemical Identifiers

CAS 150828-96-9
Molecular Formula C25H30N2O6
Molecular Weight (g/mol) 454.523
MDL Number MFCD00080279
InChI Key YEBWACZYMHWWEK-NRFANRHFSA-N
Synonym boc-orn fmoc-oh, a-fmoc-n, a-boc-l-ornithine, boc-orn fmoc-oh tlc, nalpha-tert-butoxycarbonyl-ndelta-9-fluorenylmethoxycarbonyl-l-ornithine, s-5-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-l-orn fmoc-oh, n-alpha-t-butyloxycarbonyl-n-gamma-9-fluorenylmethyloxycarbonyl-l-ornithine, n-boc-n'-fmoc-l-ornithine, n-a-boc-n-d-fmoc-l-ornithine, n, a-boc-n, a-fmoc-l-ornithine, n
PubChem CID 16211325
IUPAC Name (2S)-5-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH62869MD View Documents Alfa Aesar™
H62869MD
250mg Each for $28.84
 
AAH6286903 View Documents Alfa Aesar™
H6286903
1g Each for $75.11
 
AAH6286906 View Documents Alfa Aesar™
H6286906
5g Each for $300.72
 
Specifications

Chemical Identifiers

150828-96-9
454.523
YEBWACZYMHWWEK-NRFANRHFSA-N
16211325
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O
C25H30N2O6
MFCD00080279
boc-orn fmoc-oh, a-fmoc-n, a-boc-l-ornithine, boc-orn fmoc-oh tlc, nalpha-tert-butoxycarbonyl-ndelta-9-fluorenylmethoxycarbonyl-l-ornithine, s-5-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-l-orn fmoc-oh, n-alpha-t-butyloxycarbonyl-n-gamma-9-fluorenylmethyloxycarbonyl-l-ornithine, n-boc-n'-fmoc-l-ornithine, n-a-boc-n-d-fmoc-l-ornithine, n, a-boc-n, a-fmoc-l-ornithine, n
(2S)-5-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid

Specifications

150828-96-9
MFCD00080279
boc-orn fmoc-oh, a-fmoc-n, a-boc-l-ornithine, boc-orn fmoc-oh tlc, nalpha-tert-butoxycarbonyl-ndelta-9-fluorenylmethoxycarbonyl-l-ornithine, s-5-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino pentanoic acid, boc-l-orn fmoc-oh, n-alpha-t-butyloxycarbonyl-n-gamma-9-fluorenylmethyloxycarbonyl-l-ornithine, n-boc-n'-fmoc-l-ornithine, n-a-boc-n-d-fmoc-l-ornithine, n, a-boc-n, a-fmoc-l-ornithine, n
CC(C)(C)OC(=O)NC(CCCNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O
454.523
454.52
150°C to 154°C
Nalpha-Boc-Ndelta-Fmoc-L-ornithine
C25H30N2O6
8091167
YEBWACZYMHWWEK-NRFANRHFSA-N
(2S)-5-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
16211325
95%
250mg