Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Boc-L-ornithine, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Boc-L-ornithine
CAS 21887-64-9
Formula Weight 232.28
MDL Number MFCD00076970
Beilstein 4313353
Synonym Boc-Orn-OH; (S)-5-amino-2-(Boc-amino)pentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6201403

  SDS
alfa aesar™
H6201403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6201406

  SDS
alfa aesar™
H6201406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6201414

  SDS
alfa aesar™
H6201414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.28
Melting Point 207° to 208°C
Chemical Name or Material Nalpha-Boc-L-ornithine
Physical Form Powder
Assay 95%
MDL Number MFCD00076970
CAS 21887-64-9
Beilstein 4313353
Synonym Boc-Orn-OH; (S)-5-amino-2-(Boc-amino)pentanoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Color White
Quantity 1g
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.