missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nalpha-Boc-L-arginine hydrochloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H6361106

 View more versions of this product

Catalog No. AAH6361106



Description

Description

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Nalpha-Boc-L-arginine hydrochloride
White
MFCD00065552
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-d-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932
HDELGKMVZYHPPB-RZUXQYDTNA-N
(2S)-5-(diaminomethylideneamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid;hydrate;hydrochloride
2729004
98%
Powder
35897-34-8
C11H23ClN4O4
3658939
Slightly soluble in water.
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O
310.78
310.78
5 g

Chemical Identifiers

35897-34-8
310.78
HDELGKMVZYHPPB-RZUXQYDTNA-N
2729004
C11H23ClN4O4
MFCD00065552
boc-l-arg-oh.hcl.h2o, n-boc-l-arginine hydrochloride monohydrate, n-boc-l-arginine hydrochloride monohydrate, boc-d-arg-oh hcl h2o, boc-d-arg-oh hcl h2o, nalpha-tert-butoxycarbonyl-l-arginine hydrochloride monohydrate, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, 2s-2-tert-butoxycarbonyl amino-5-carbamimidamidopentanoic acid hydrate hydrochloride, n-, a-boc-d-arginine hydrochloride monohydrate, pubchem14932
Cl.CC(C)(C)OC(=O)N[C@@H](CCCNC(N)=N)C(O)=O
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only