missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H6360903
Description
Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsNα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.
Solubility
Slightly soluble in water and dimethyl sulfoxide.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine | |
C31H38N4O6S | |
fmoc-lys biotin-oh, fmoc-lys biotinyl-oh, fmoc-lys biotinyl-oh, nalpha-fmoc-nepsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, fmoc biocytin, fmoc biocytin, fmoc-lys biotin, fmoc-lys biotin, n, a-fmoc-biocytin | |
OFIBQNGDYNGUEZ-CVKYCUBCNA-N | |
2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-6-(5-{2-oxo-hexahydro-1H-thieno[3,4-d]imidazol-4-yl}pentanamido)hexanoic acid | |
11039242 | |
95% | |
Solid |
146987-10-2 | |
MFCD00270741 | |
Slightly soluble in water and dimethyl sulfoxide. | |
[H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2 | |
594.73 | |
594.72 | |
1 g |
Chemical Identifiers
146987-10-2 | |
594.73 | |
OFIBQNGDYNGUEZ-CVKYCUBCNA-N | |
11039242 |
C31H38N4O6S | |
MFCD00270741 | |
fmoc-lys biotin-oh, fmoc-lys biotinyl-oh, fmoc-lys biotinyl-oh, nalpha-fmoc-nepsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, fmoc biocytin, fmoc biocytin, fmoc-lys biotin, fmoc-lys biotin, n, a-fmoc-biocytin | |
[H][C@]12CS[C@@H](CCCCC(=O)NCCCC[C@H](NC(=O)OCC3C4=CC=CC=C4C4=CC=CC=C34)C(O)=O)[C@@]1([H])NC(=O)N2 |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only