missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine, 95%

Manufacturer:  Alfa Aesar™ H63609MD

 View more versions of this product

Catalog No. AAH63609MD



Description

Description

Applications
Nα-Fmoc-Nε-biotinyl-L-lysine is useful in the synthesis of site-specific biotinylated probes.

Solubility
Slightly soluble in water and dimethyl sulfoxide.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Nalpha-Biotinyl-Nepsilon-Fmoc-L-lysine
C31H38N4O6S
fmoc-lys biotin-oh, fmoc-l-lys biotinyl-oh, fmoc-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamido hexanoic acid, fmoc biocytin, fmoc-biocytin, fmoc-lys biotin, n, a-fmoc-biocytin
C1C2C(C(S1)CCCCC(=O)NCCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)NC(=O)N2
594.727
594.72
95%
146987-10-2
MFCD00270741
OFIBQNGDYNGUEZ-OBXRUURASA-N
(2S)-6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
11039242
Solid
250mg

Chemical Identifiers

146987-10-2
594.727
OFIBQNGDYNGUEZ-OBXRUURASA-N
11039242
C1C2C(C(S1)CCCCC(=O)NCCCCC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35)NC(=O)N2
C31H38N4O6S
MFCD00270741
fmoc-lys biotin-oh, fmoc-l-lys biotinyl-oh, fmoc-lys biotinyl-oh, fmoc-n-epsilon-biotinyl-l-lysine, nalpha-fmoc-nepsilon-biotinyl-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-5-3as,4s,6ar-2-oxohexahydro-1h-thieno 3,4-d imidazol-4-yl pentanamido hexanoic acid, fmoc biocytin, fmoc-biocytin, fmoc-lys biotin, n, a-fmoc-biocytin
(2S)-6-[5-[(3aS,4S,6aR)-2-oxo-1,3,3a,4,6,6a-hexahydrothieno[3,4-d]imidazol-4-yl]pentanoylamino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
SDS Documents