Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-tyrosine dicyclohexylammonium salt, 98%

Chemical Identifiers

Chemical Name or Material Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-tyrosine dicyclohexylammonium salt
CAS 198828-72-7
Formula Weight 552.74
MDL Number MFCD00065702
Synonym Z-D-Tyr(tBu)-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6304203

  SDS
alfa aesar™
H6304203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6304206

  SDS
alfa aesar™
H6304206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 552.74
Chemical Name or Material Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-tyrosine dicyclohexylammonium salt
Physical Form Solid
Assay 98%
MDL Number MFCD00065702
CAS 198828-72-7
Synonym Z-D-Tyr(tBu)-OH.DCHA
Quantity 1g
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C