missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-tyrosine dicyclohexylammonium salt, 98%

Manufacturer:  Alfa Aesar™ H6304206

 View more versions of this product

Catalog No. AAH6304206



Specifications

Specifications

Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-tyrosine dicyclohexylammonium salt
C33H48N2O5
z-d-tyr tbu-oh.dcha, z-d-tyr tbu-oh dcha, dicyclohexylamine r-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoate, z-d-tyr tbu-ohcha, z-d-tyr but-oh dcha, z-d-tyr tbu-oh?cha, n-alpha-benzyloxycarbonyl-o-t-butyl-d-tyrosine dicyclohexylamine, 2r-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid; dicha, n-benzyloxy carbonyl-o-tert-butyl-d-tyrosine-n-cyclohexylcyclohexanamine 1/1, n^a-benzyloxycarbonyl-o-tert-butyl-d-tyrosine dicyclohexylammonium salt
CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2.C1CCC(CC1)NC2CCCCC2
552.756
552.74
98%
198828-72-7
MFCD00065702
FDNJRKLIHBJXIR-GMUIIQOCSA-N
N-cyclohexylcyclohexanamine;(2R)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]-2-(phenylmethoxycarbonylamino)propanoic acid
53229926
Solid
5g

Chemical Identifiers

198828-72-7
552.756
FDNJRKLIHBJXIR-GMUIIQOCSA-N
53229926
CC(C)(C)OC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2=CC=CC=C2.C1CCC(CC1)NC2CCCCC2
C33H48N2O5
MFCD00065702
z-d-tyr tbu-oh.dcha, z-d-tyr tbu-oh dcha, dicyclohexylamine r-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoate, z-d-tyr tbu-ohcha, z-d-tyr but-oh dcha, z-d-tyr tbu-oh?cha, n-alpha-benzyloxycarbonyl-o-t-butyl-d-tyrosine dicyclohexylamine, 2r-2-benzyloxy carbonyl amino-3-4-tert-butoxy phenyl propanoic acid; dicha, n-benzyloxy carbonyl-o-tert-butyl-d-tyrosine-n-cyclohexylcyclohexanamine 1/1, n^a-benzyloxycarbonyl-o-tert-butyl-d-tyrosine dicyclohexylammonium salt
N-cyclohexylcyclohexanamine;(2R)-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]-2-(phenylmethoxycarbonylamino)propanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents