missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-serine, 98%

$17.59 - $143.11

Chemical Identifiers

CAS 65806-90-8
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00077038
InChI Key TXDGEONUWGOCJG-GFCCVEGCSA-N
Synonym z-d-ser tbu-oh, cbz-o-tert-butyl-d-serine, r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-cbz-o-tert-butyl-d-serine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, ambotzzaa1152, n-alpha-benzyloxycarbonyl-o-t-butyl-d-serine, n-alpha-carbobenzoxy-d-serine t-butyl ether, r-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-d-serine
PubChem CID 1715619
IUPAC Name (2R)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63728MD View Documents Alfa Aesar™
H63728MD
250mg Each for $17.59
 
AAH6372803 View Documents Alfa Aesar™
H6372803
1g Each for $36.39
 
AAH6372806 View Documents Alfa Aesar™
H6372806
5g Each for $143.11
 
Specifications

Chemical Identifiers

65806-90-8
295.335
TXDGEONUWGOCJG-GFCCVEGCSA-N
1715619
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C15H21NO5
MFCD00077038
z-d-ser tbu-oh, cbz-o-tert-butyl-d-serine, r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-cbz-o-tert-butyl-d-serine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, ambotzzaa1152, n-alpha-benzyloxycarbonyl-o-t-butyl-d-serine, n-alpha-carbobenzoxy-d-serine t-butyl ether, r-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-d-serine
(2R)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

65806-90-8
MFCD00077038
TXDGEONUWGOCJG-GFCCVEGCSA-N
(2R)-3-[(2-methylpropan-2-yl)oxy]-2-(phenylmethoxycarbonylamino)propanoic acid
1715619
Powder
White
Nalpha-Benzyloxycarbonyl-O-tert-butyl-D-serine
C15H21NO5
z-d-ser tbu-oh, cbz-o-tert-butyl-d-serine, r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, n-cbz-o-tert-butyl-d-serine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxy propanoic acid, ambotzzaa1152, n-alpha-benzyloxycarbonyl-o-t-butyl-d-serine, n-alpha-carbobenzoxy-d-serine t-butyl ether, r-2-benzyloxycarbonylamino-3-tert-butoxypropanoic acid, o-1,1-dimethylethyl-n-phenylmethoxy carbonyl-d-serine
CC(C)(C)OCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
295.335
295.33
98%
250mg
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C