missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine, 98%

$70.39 - $306.97

Chemical Identifiers

CAS 2304-98-5
Molecular Formula C14H19N5O6
Molecular Weight (g/mol) 353.335
MDL Number MFCD00038092
InChI Key BZPCSFNCKORLQG-NSHDSACASA-N
Synonym 2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, cbz-arg no2-oh, cbz-l-arg no2-oh, l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl, n-alpha-z-n-omega-nitro-l-arginine, n-cbz-n-nitro-l-arginine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine, s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid, z-arg no2-oh, z-l-arg no2-oh
PubChem CID 12695023
SMILES C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6339206
View Documents
Alfa Aesar™
H6339206
5g Each for $70.39
 
AAH6339214
View Documents
Alfa Aesar™
H6339214
25g Each for $306.97
 
Specifications

Chemical Identifiers

2304-98-5
353.335
BZPCSFNCKORLQG-NSHDSACASA-N
12695023
C14H19N5O6
MFCD00038092
2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, cbz-arg no2-oh, cbz-l-arg no2-oh, l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl, n-alpha-z-n-omega-nitro-l-arginine, n-cbz-n-nitro-l-arginine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine, s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid, z-arg no2-oh, z-l-arg no2-oh
C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O

Specifications

2304-98-5
MFCD00038092
BZPCSFNCKORLQG-NSHDSACASA-N
353.335
353.33
98%
Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine
C14H19N5O6
2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, cbz-arg no2-oh, cbz-l-arg no2-oh, l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl, n-alpha-z-n-omega-nitro-l-arginine, n-cbz-n-nitro-l-arginine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine, s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid, z-arg no2-oh, z-l-arg no2-oh
C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
12695023
Solid
5g
Safety and Handling

Safety and Handling

EINECSNumber : 219-970-0

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold