missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine, 98%

Manufacturer:  Alfa Aesar™ H6339214

 View more versions of this product

Catalog No. AAH6339214



Specifications

Specifications

Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine
C14H19N5O6
z-arg no2-oh, z-l-arg no2-oh, n-cbz-n-nitro-l-arginine, cbz-l-arg no2-oh, s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid, l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl, cbz-arg no2-oh, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine, 2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, n-alpha-z-n-omega-nitro-l-arginine
C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
12695023
Solid
25g
2304-98-5
MFCD00038092
BZPCSFNCKORLQG-NSHDSACASA-N
353.335
353.33
98%

Chemical Identifiers

2304-98-5
353.335
BZPCSFNCKORLQG-NSHDSACASA-N
12695023
C14H19N5O6
MFCD00038092
z-arg no2-oh, z-l-arg no2-oh, n-cbz-n-nitro-l-arginine, cbz-l-arg no2-oh, s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid, l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl, cbz-arg no2-oh, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine, 2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid, n-alpha-z-n-omega-nitro-l-arginine
C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents