Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Nomega-(4-methoxybenzenesulfonyl)-L-arginine dicyclohexylammonium salt, 95%

Chemical Identifiers

Chemical Name or Material Nalpha-Benzyloxycarbonyl-Nomega-(4-methoxybenzenesulfonyl)-L-arginine dicyclohexylammonium salt
CAS 58810-11-0
Formula Weight 659.84
MDL Number MFCD00237326
Synonym Z-Arg(Mbs)-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6624003

 
alfa aesar™
H6624003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6624006

 
alfa aesar™
H6624006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66240MD

 
alfa aesar™
H66240MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 659.84
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Nomega-(4-methoxybenzenesulfonyl)-L-arginine dicyclohexylammonium salt
CAS 58810-11-0
Assay 95%
MDL Number MFCD00237326
Physical Form Solid
Synonym Z-Arg(Mbs)-OH.DCHA
Storage Note 1 -30° to -10°C
Quantity 1g
Safety and Handling

-30° to -10°C