Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Nepsilon-Boc-L-lysine 4-nitrophenyl ester, 98%

Chemical Identifiers

Chemical Name or Material Nalpha-Benzyloxycarbonyl-Nepsilon-Boc-L-lysine 4-nitrophenyl ester
CAS 2212-69-3
Formula Weight 501.54
MDL Number MFCD00076963
Synonym Z-Lys(Boc)-ONP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6606003

 
alfa aesar™
H6606003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6606006

 
alfa aesar™
H6606006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66060MD

 
alfa aesar™
H66060MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 501.54
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Nepsilon-Boc-L-lysine 4-nitrophenyl ester
CAS 2212-69-3
MDL Number MFCD00076963
Synonym Z-Lys(Boc)-ONP
Physical Form Solid
Storage Note 1 2° to 8°C
Quantity 1g
Assay 98%
Safety and Handling

2° to 8°C