Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-L-ornithine, 98%

$44.59 - $44.59

Chemical Identifiers

Chemical Name or Material Nalpha-Benzyloxycarbonyl-L-ornithine
CAS 2640-58-6
Formula Weight 266.29
MDL Number MFCD00038200
Synonym Z-Orn-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6305206

  SDS
alfa aesar™
H6305206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6305203

  SDS
alfa aesar™
H6305203
1g Each for $44.59
Description & Specifications

Specifications

Formula Weight 266.29
MDL Number MFCD00038200
Synonym Z-Orn-OH
Physical Form Powder
Color White
CAS 2640-58-6
Assay 98%
Storage Note 1 2° to 8°C
Chemical Name or Material Nalpha-Benzyloxycarbonyl-L-ornithine
Quantity 5g

Applications
Nalpha-Benzyloxycarbonyl-L-ornithine is widely used as pharmaceutical intermediate.

Solubility
Soluble in 0.1N NaOG (1 mg/ml-clear, colorless solution). Slightly soluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.

Safety and Handling

2° to 8°C