missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-DL-histidine, 98%

$106.26 - $422.54

Chemical Identifiers

CAS 19728-57-5
Molecular Formula C14H15N3O4
Molecular Weight (g/mol) 289.291
MDL Number MFCD00037852
InChI Key WCOJOHPAKJFUDF-UHFFFAOYSA-N
Synonym n-cbz-dl-histidine, z-dl-his-oh, cbz-dl-histidine, 2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid, carbobenzoxy-dl-histidine, 2-benzyloxy carbonyl amino-3-3h-imidazol-4-yl propanoic acid, n-benzyloxy carbonyl histidine, acmc-209u4b, n-carbobenzoxy-dl-histidine, .alpha.-n-cbz-dl-histidine
PubChem CID 85793
IUPAC Name 3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6615206 View Documents Alfa Aesar™
H6615206
5g Each for $106.26
 
AAH6615214 View Documents Alfa Aesar™
H6615214
25g Each for $422.54
 
Specifications

Chemical Identifiers

19728-57-5
289.291
WCOJOHPAKJFUDF-UHFFFAOYSA-N
85793
C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O
C14H15N3O4
MFCD00037852
n-cbz-dl-histidine, z-dl-his-oh, cbz-dl-histidine, 2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid, carbobenzoxy-dl-histidine, 2-benzyloxy carbonyl amino-3-3h-imidazol-4-yl propanoic acid, n-benzyloxy carbonyl histidine, acmc-209u4b, n-carbobenzoxy-dl-histidine, .alpha.-n-cbz-dl-histidine
3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

19728-57-5
MFCD00037852
WCOJOHPAKJFUDF-UHFFFAOYSA-N
3-(1H-imidazol-5-yl)-2-(phenylmethoxycarbonylamino)propanoic acid
85793
Powder
White
Nalpha-Benzyloxycarbonyl-DL-histidine
C14H15N3O4
n-cbz-dl-histidine, z-dl-his-oh, cbz-dl-histidine, 2-benzyloxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid, carbobenzoxy-dl-histidine, 2-benzyloxy carbonyl amino-3-3h-imidazol-4-yl propanoic acid, n-benzyloxy carbonyl histidine, acmc-209u4b, n-carbobenzoxy-dl-histidine, .alpha.-n-cbz-dl-histidine
C1=CC=C(C=C1)COC(=O)NC(CC2=CN=CN2)C(=O)O
289.291
289.29
98%
5g
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C