missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-D-lysine, 95%

$72.47 - $302.26

Chemical Identifiers

CAS 70671-54-4
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.324
MDL Number MFCD00067713
InChI Key OJTJKAUNOLVMDX-GFCCVEGCSA-N
Synonym z-d-lys-oh, a-cbz-d-lysine, cbz-d-lysine, nalpha-carbobenzoxy-d-lysine, nalpha-cbz-d-lysine, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, n-alpha-benzyloxycarbonyl-d-lysine, r-6-amino-2-benzyloxycarbonylamino hexanoic acid, n-cbz-d-lysine, nalpha-z-d-lysine, n
PubChem CID 7017997
IUPAC Name (2R)-6-amino-2-(phenylmethoxycarbonylamino)hexanoic acid
SMILES C1=CC=C(C=C1)COC(=O)NC(CCCCN)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6564003 View Documents Alfa Aesar™
H6564003
1g Each for $72.47
 
AAH6564006 View Documents Alfa Aesar™
H6564006
5g Each for $302.26
 
Specifications

Chemical Identifiers

70671-54-4
280.324
OJTJKAUNOLVMDX-GFCCVEGCSA-N
7017997
C1=CC=C(C=C1)COC(=O)NC(CCCCN)C(=O)O
C14H20N2O4
MFCD00067713
z-d-lys-oh, a-cbz-d-lysine, cbz-d-lysine, nalpha-carbobenzoxy-d-lysine, nalpha-cbz-d-lysine, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, n-alpha-benzyloxycarbonyl-d-lysine, r-6-amino-2-benzyloxycarbonylamino hexanoic acid, n-cbz-d-lysine, nalpha-z-d-lysine, n
(2R)-6-amino-2-(phenylmethoxycarbonylamino)hexanoic acid

Specifications

70671-54-4
MFCD00067713
z-d-lys-oh, a-cbz-d-lysine, cbz-d-lysine, nalpha-carbobenzoxy-d-lysine, nalpha-cbz-d-lysine, r-6-amino-2-benzyloxy carbonyl amino hexanoic acid, n-alpha-benzyloxycarbonyl-d-lysine, r-6-amino-2-benzyloxycarbonylamino hexanoic acid, n-cbz-d-lysine, nalpha-z-d-lysine, n
C1=CC=C(C=C1)COC(=O)NC(CCCCN)C(=O)O
280.324
280.32
95%
1g
C14H20N2O4
5290234
OJTJKAUNOLVMDX-GFCCVEGCSA-N
(2R)-6-amino-2-(phenylmethoxycarbonylamino)hexanoic acid
7017997
Crystalline
White
Nalpha-Benzyloxycarbonyl-D-lysine