missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-3-(Boc-amino)-D-alanine, 96%

$43.49 - $515.07

Chemical Identifiers

CAS 62234-36-0
Molecular Formula C16H22N2O6
Molecular Weight (g/mol) 338.36
MDL Number MFCD01318749
InChI Key WJKGPJRAGHSOLM-GFCCVEGCSA-N
Synonym z-d-dap boc-oh, d-n-cbz-3-n-boc-amino-alanine, r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, n-a-z-n-beta-boc-d-2,3-diaminopropionic acid, n-alpha-benzyloxycarbonyl-n-beta-t-butyloxycarbonyl-d-2,3-diaminopropionic acid, r-2-benzyloxycarbonylamino-3-tert-butoxycarbonylamino-propionic acid, 2r-2-benzyloxycarbonyl amino-3-tert-butoxycarbonyl amino propionic acid, 3-1,1-dimethylethoxy carbonyl amino-n-phenylmethoxy carbonyl-d-alanine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, 2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid
PubChem CID 7006678
IUPAC Name (2R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)propanoic acid
SMILES CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6630103 View Documents Alfa Aesar™
H6630103
1g Each for $129.08
 
AAH6630106 View Documents Alfa Aesar™
H6630106
5g Each for $515.07
 
AAH66301MD View Documents Alfa Aesar™
H66301MD
250mg Each for $43.49
 
Specifications

Chemical Identifiers

62234-36-0
338.36
WJKGPJRAGHSOLM-GFCCVEGCSA-N
7006678
CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
C16H22N2O6
MFCD01318749
z-d-dap boc-oh, d-n-cbz-3-n-boc-amino-alanine, r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, n-a-z-n-beta-boc-d-2,3-diaminopropionic acid, n-alpha-benzyloxycarbonyl-n-beta-t-butyloxycarbonyl-d-2,3-diaminopropionic acid, r-2-benzyloxycarbonylamino-3-tert-butoxycarbonylamino-propionic acid, 2r-2-benzyloxycarbonyl amino-3-tert-butoxycarbonyl amino propionic acid, 3-1,1-dimethylethoxy carbonyl amino-n-phenylmethoxy carbonyl-d-alanine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, 2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid
(2R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)propanoic acid

Specifications

62234-36-0
MFCD01318749
WJKGPJRAGHSOLM-GFCCVEGCSA-N
(2R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)propanoic acid
7006678
Solid
1g
C16H22N2O6
z-d-dap boc-oh, d-n-cbz-3-n-boc-amino-alanine, r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, n-a-z-n-beta-boc-d-2,3-diaminopropionic acid, n-alpha-benzyloxycarbonyl-n-beta-t-butyloxycarbonyl-d-2,3-diaminopropionic acid, r-2-benzyloxycarbonylamino-3-tert-butoxycarbonylamino-propionic acid, 2r-2-benzyloxycarbonyl amino-3-tert-butoxycarbonyl amino propionic acid, 3-1,1-dimethylethoxy carbonyl amino-n-phenylmethoxy carbonyl-d-alanine, 2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid, 2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid
CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
338.36
338.36
96%
Nalpha-Benzyloxycarbonyl-3-(Boc-amino)-D-alanine
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C