missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Benzoyl-L-ornithine, 98%

$25.59 - $297.64

Chemical Identifiers

CAS 17966-71-1
Molecular Formula C12H16N2O3
Molecular Weight (g/mol) 236.271
MDL Number MFCD00153452
InChI Key PKONOCQSEOHHJW-JTQLQIEISA-N
Synonym n2-benzoyl-l-ornithine, bz-orn-oh, n-benzoyl-ornithine, s-5-amino-2-benzamidopentanoic acid, 2s-5-amino-2-phenylformamido pentanoic acid, benzoylornithin, ambotzbaa0043, l-ornithine, n2-benzoyl, n-alpha-benzoyl-l-ornithine
PubChem CID 7016391
IUPAC Name (2S)-5-amino-2-benzamidopentanoic acid
SMILES C1=CC=C(C=C1)C(=O)NC(CCCN)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6615603 View Documents Alfa Aesar™
H6615603
1g Each for $75.11
 
AAH6615606 View Documents Alfa Aesar™
H6615606
5g Each for $297.64
 
AAH66156MD View Documents Alfa Aesar™
H66156MD
250mg Each for $25.59
 
Specifications

Chemical Identifiers

17966-71-1
236.271
PKONOCQSEOHHJW-JTQLQIEISA-N
7016391
C1=CC=C(C=C1)C(=O)NC(CCCN)C(=O)O
C12H16N2O3
MFCD00153452
n2-benzoyl-l-ornithine, bz-orn-oh, n-benzoyl-ornithine, s-5-amino-2-benzamidopentanoic acid, 2s-5-amino-2-phenylformamido pentanoic acid, benzoylornithin, ambotzbaa0043, l-ornithine, n2-benzoyl, n-alpha-benzoyl-l-ornithine
(2S)-5-amino-2-benzamidopentanoic acid

Specifications

17966-71-1
MFCD00153452
PKONOCQSEOHHJW-JTQLQIEISA-N
(2S)-5-amino-2-benzamidopentanoic acid
7016391
Powder
White
Nalpha-Benzoyl-L-ornithine
C12H16N2O3
n2-benzoyl-l-ornithine, bz-orn-oh, n-benzoyl-ornithine, s-5-amino-2-benzamidopentanoic acid, 2s-5-amino-2-phenylformamido pentanoic acid, benzoylornithin, ambotzbaa0043, l-ornithine, n2-benzoyl, n-alpha-benzoyl-l-ornithine
C1=CC=C(C=C1)C(=O)NC(CCCN)C(=O)O
236.271
236.27
98%
1g
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C