Please login to your online account to display your discounted pricing

Nalpha-Benzoyl-L-arginine ethyleester hydrochloride, ≥99%, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: fisher scientific acros  105700100

 View more versions of this product

Catalog No. AC105700100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Powder, Crystals or Granules
Formula Weight 342.83
Quantity 10g
Color White to slightly yellow
Beilstein 09, III, 1232
Melting Point 129° to 136°C
Chemical Name or Material Nα-Benzoyl-L-arginine ethyl ester hydrochloride
Individual Impurity 99.0% min. (TLC)
Molecular Formula C15H22N4O3·HCl
Linear Formula H2NC(=NH)NH(CH2)3CH(NHCOC6H5)CO2C2H5·HCl
Packaging Glass bottle
Synonym BAEE
Assay Percent Range 99+%
Infrared Spectrum Authentic
MDL Number MFCD00012579
CAS 272320
CAS Min % 99.0
Specific Rotation −15.0° to −19.0° (24°C, 589nm) (c=2, water)
CAS Max % 100.0
Appearance of Solution (50mg/mLin water) clear colorless

Specifications

Chemical Name or Material Nα-Benzoyl-L-arginine ethyl ester hydrochloride
CAS 272320
CAS Min % 99.0
CAS Max % 100.0
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C15H22N4O3·HCl
Linear Formula H2NC(=NH)NH(CH2)3CH(NHCOC6H5)CO2C2H5·HCl
Formula Weight 342.83
MDL Number MFCD00012579
Synonym BAEE