missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalpha-Acetyl-Nepsilon-Boc-L-lysine, 95%

$95.15 - $629.19

Chemical Identifiers

CAS 23500-04-1
Molecular Formula C13H24N2O5
Molecular Weight (g/mol) 288.344
MDL Number MFCD00236761
InChI Key NZAMQYCOJSWUAO-JTQLQIEISA-N
Synonym ac-lys boc-oh, s-2-acetamido-6-tert-butoxycarbonyl amino hexanoic acid, acetyl-nepsilon-boc-l-lysine, 2s-6-tert-butoxycarbonyl amino-2-acetamidohexanoic acid, n~2~-acetyl-n~6~-tert-butoxycarbonyl-l-lysine, s-2-acetamido-6-tert-butoxycarbonyl amino hexanoicacid
PubChem CID 7019610
IUPAC Name (2S)-2-acetamido-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(=O)NC(CCCCNC(=O)OC(C)(C)C)C(=O)O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6617206 View Documents Alfa Aesar™
H6617206
5g Each for $95.15
 
AAH6617214 View Documents Alfa Aesar™
H6617214
25g Each for $237.49
 
AAH6617222 View Documents Alfa Aesar™
H6617222
100g Each for $629.19
 
Specifications

Chemical Identifiers

23500-04-1
288.344
NZAMQYCOJSWUAO-JTQLQIEISA-N
7019610
CC(=O)NC(CCCCNC(=O)OC(C)(C)C)C(=O)O
C13H24N2O5
MFCD00236761
ac-lys boc-oh, s-2-acetamido-6-tert-butoxycarbonyl amino hexanoic acid, acetyl-nepsilon-boc-l-lysine, 2s-6-tert-butoxycarbonyl amino-2-acetamidohexanoic acid, n~2~-acetyl-n~6~-tert-butoxycarbonyl-l-lysine, s-2-acetamido-6-tert-butoxycarbonyl amino hexanoicacid
(2S)-2-acetamido-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid

Specifications

23500-04-1
MFCD00236761
NZAMQYCOJSWUAO-JTQLQIEISA-N
(2S)-2-acetamido-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
7019610
Powder
White
Nalpha-Acetyl-Nepsilon-Boc-L-lysine
C13H24N2O5
ac-lys boc-oh, s-2-acetamido-6-tert-butoxycarbonyl amino hexanoic acid, acetyl-nepsilon-boc-l-lysine, 2s-6-tert-butoxycarbonyl amino-2-acetamidohexanoic acid, n~2~-acetyl-n~6~-tert-butoxycarbonyl-l-lysine, s-2-acetamido-6-tert-butoxycarbonyl amino hexanoicacid
CC(=O)NC(CCCCNC(=O)OC(C)(C)C)C(=O)O
288.344
288.34
95%
5g
Safety and Handling

Safety and Handling

storageNote1 : -30° to -10°C