Please login to your online account to display your discounted pricing

Naloxone benzoylhydrazone, Tocris

Full κ agonist, partial μ and δ agonist and antagonizes agonist-induced activation of NOP

$146.74 - $618.87

Chemical Identifiers

Chemical Name or Material Naloxone benzoylhydrazone
Assay Percent Range >99%
Molecular Formula C26H27N3O4
Formula Weight 445.52
Synonym [(5α)-4, 5-Epoxy-3, 14-dihydroxy-17-(2-propenyl)morphinan-6-ylidene] benzoic acid hydrazide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

141950

 
r & d systems
RDS141950
50mg Each for $618.87

141910

 
r & d systems
RDS141910
10mg Each for $146.74
Description & Specifications

Specifications

Formula Weight 445.52
Quantity 50mg
CAS 119630-94-3
Molecular Formula C26H27N3O4
Chemical Name or Material Naloxone benzoylhydrazone
Assay Percent Range >99%
Synonym [(5α)-4, 5-Epoxy-3, 14-dihydroxy-17-(2-propenyl)morphinan-6-ylidene] benzoic acid hydrazide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C