Please login to your online account to display your discounted pricing

Naloxonazine dihydrochloride, Tocris


Selective μ1 antagonist

Quantity 50mg

Manufacturer: r & d systems  RDS059150

 View more versions of this product

Catalog No. 059150

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 723.69
Chemical Name or Material Naloxonazine dihydrochloride
Assay Percent Range >98%
Molecular Formula C38H42N4O6·2HCl
CAS 880759-65-9
Synonym Bis-[5-α-4, 5-Epoxy-3, 14-dihydroxy-17-(2-propenyl)-morphinan-6-ylidene] hydrazine dihydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material Naloxonazine dihydrochloride
Assay Percent Range >98%
Molecular Formula C38H42N4O6·2HCl
Formula Weight 723.69
Synonym Bis-[5-α-4, 5-Epoxy-3, 14-dihydroxy-17-(2-propenyl)-morphinan-6-ylidene] hydrazine dihydrochloride
Accessories

Store at Room Temperature