Please login to your online account to display your discounted pricing

Naloxonazine dihydrochloride, Tocris

Selective μ1 antagonist

$129.05 - $546.19

Chemical Identifiers

Chemical Name or Material Naloxonazine dihydrochloride
Assay Percent Range >98%
Molecular Formula C38H42N4O6·2HCl
Formula Weight 723.69
Synonym Bis-[5-α-4, 5-Epoxy-3, 14-dihydroxy-17-(2-propenyl)-morphinan-6-ylidene] hydrazine dihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

059110

 
r & d systems
RDS059110
10mg Each for $129.05

059150

 
r & d systems
RDS059150
50mg Each for $546.19
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 723.69
Chemical Name or Material Naloxonazine dihydrochloride
Assay Percent Range >98%
Molecular Formula C38H42N4O6·2HCl
CAS 880759-65-9
Synonym Bis-[5-α-4, 5-Epoxy-3, 14-dihydroxy-17-(2-propenyl)-morphinan-6-ylidene] hydrazine dihydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature