Please login to your online account to display your discounted pricing

Alfa Aesar™ Nalidixic acid, 50mg/mL in water, sterile-filtered

Chemical Identifiers

Chemical Name or Material Nalidixic acid
CAS 389-08-2
Name Note 50mg/mL in water, sterile-filtered
MDL Number MFCD00006884
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ671948EQ

 
alfa aesar™
J671948EQ
5 x 1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67194XF

 
alfa aesar™
J67194XF
1mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Nalidixic acid
CAS 389-08-2
Physical Form Liquid
MDL Number MFCD00006884
Name Note 50mg/mL in water, sterile-filtered
Storage Note 1 -30° to -10°C
RTECS Number QN2885000
Quantity 5 x 1mL

Applications
Nalidixic acid is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. It is used to modulate (inhibit) bacterial DNA gyrase-dependent processes such as DNA polymerization, (ATP-dependent)t DNA supercoiling, and chromosome fragmentation.

Solubility
Soluble in chloroform (50 mg/ml), ethanol (0.9 mg/ml) at 23°C, methanol (1.3 mg/ml) at 23°C, toluene (1.6 mg/ml) at 23°C, DMSO (≥ 6 mg/ml), and solution of carbonates. Fully miscible in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.

Safety and Handling

-30° to -10°C