Thermo Scientific™ Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge
Obtain full blank port pressure with only two strokes when using the Thermo Scientific™ Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge.
$770.79 - $770.79
Products ${productFamilyLength}
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
13-874-614A Thermo Scientific™
N61330010
Vacuum Pump Each for $770.79
Case of 4 Each for $3,083.16
Specifications
Description
Obtain full blank port pressure with only two strokes when using the Thermo Scientific™ Nalgene™ Zinc/Aluminum Hand-Operated Vacuum Pump with Gauge, a multi-purpose self-lubricating pump featuring outstanding efficiency. Cast in corrosion-resistant zinc/aluminum and easily disassembled for repair or replacement of internal parts, this hand pump quickly and easily attains a vacuum of 25″ (635mm) Hg or a pressure of 30 psi (35kPa).
  • Designed with ergonomic handle to make pumping easier on the hands
  • Corrosion resistant and repairable for long life in the lab
  • Pumping rate of 15cc per stroke
  • Easy-flip lever to switch from vacuum to pressure.
  • Gauge calibrated in inches of Mercury and in kPa
  • Attaches to 1/4″ (63.5cm) I.D. flexible tubing; includes a 2′ (0.7m) length of clear PVC tubing
  • Includes repair kit 6133-2000 for maintenance and repair of internal parts to extend hand pump life
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.