Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Wide-Mouth Opaque Amber HDPE Bottles with Closure: Bulk Pack


Store, ship and pack light-sensitive reagents in these leakproof, wide-mouthed bottles.

Manufacturer: thermo scientific™  N3121060002

 View more versions of this product

 Disclaimers

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for two minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Catalog No. 03-313-421

  • / Case of 1000 Each
 

Description & Specifications

Specifications

Closure Type Screw closure (bulk packed, unassembled)
Closure Size 28mm
Neck Finish 28mm
Mouth Wide
Height (Metric) 86mm
Shape Round
Diameter (Metric) 39mm
Sterility Nonsterile
Capacity (English) 2 oz.
Capacity (Metric) 60mL
Closure Material Polypropylene (PP)
No. per Pack 1000
Color Opaque amber
Certifications/Compliance Manufactured in compliance with ISO 13485:2003 quality requirements. Bottles and Closures meet the requirements of 21CFR177.1520 for food and beverage use, USP Classs VI. Product meets the requirements for light-resistant containers per USP latest edition
Diameter (Metric) Inner Neck 21mm
No. per Case 1000
Description Wide-Mouth Bottle
Capacity 2 oz., 60mL
Material HDPE

Store, ship and pack light-sensitive reagents in Thermo Scientific™ Nalgene™ Wide-Mouth Opaque Amber HDPE Bottles with Closure.

Features:

  • Opaque amber polypropylene closures
  • Uniform walls that resist splitting and puncturing
  • Wide mouth for easy filling and dispensing
  • Bulk packed for cost-efficiency
  • Leakproof† to prevent product loss

Ordering Information:

Bottles and closures are packed in separate polybags within the same carton.