Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Carboys with Handles
Store and transport solids and powders with the Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Carboys with Handles.
$197.52 - $277.94
Specifications
Closure Size 100-415mm
No. per Pack 1
View More Specs
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Products
Catalog Number Mfr. No. Capacity (Metric) Closure Size Price Quantity    
02-961-60A
thermo scientific™
N22340020
10L 100-415mm Each for $197.52
Case of 6 Each for $1,185.12
02-961-60C
thermo scientific™
N22340030
15L 100-415mm Each for $243.61
Case of 6 Each for $1,461.66
02-961-60E
thermo scientific™
N22340050
20L 100-415mm Each for $277.94
Case of 4 Each for $1,111.76
Description & Specifications
Specifications
Closure Size 100-415mm
No. per Pack 1
Store and transport solids and powders with the Thermo Scientific™ Nalgene™ Wide-Mouth LDPE Carboys with Handles. A convenient, large opening promotes ease of cleaning and dispensing and can accommodate overhead mixers while its wide-shoulder handles enable easy carrying and pouring, even with gloved hands. Carboy includes leakproof white PP (polypropylene) 100mm screw closures.
  • Constructed of low density polyethylene
  • Graduated to contain in 1 gal. and 5L increments for ease of use
  • Wide-mouth opening to accommodate overhead mixers and is convenient for dry materials such as solids and powders
  • Leakproof†/Not autoclavable
.