Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Centrifuge Bottles
Carry out applications such as large-volume cell harvesting, pelleting and protein purification with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Centrifuge Bottles.
$137.27 - $137.27
Products
Catalog Number Mfr. No. Max. Force Nominal Capacity Price Quantity    
05-562-50
thermo scientific™
31210250
8,000 x g 250mL Pack of 6 Each for $137.27
Case of 6 Pack for $823.62
Description & Specifications
Carry out applications such as large-volume cell harvesting, pelleting and protein purification with Thermo Scientific™ Nalgene™ Wide-Mouth HDPE Centrifuge Bottles, molded of rigid and robust high-density polyethylene. These translucent bottles have excellent chemical resistance properties and are rated for temperatures as low as −100°C.
Features:
  • For low speed centrifugation of biological and chemical samples
  • Excellent for use in refrigerated centrifuges
  • Rated for forces up to 8000 x g
  • For proper performance bottles, must be filled to at least 80% of total capacity
Requires:
When spinning 250mL bottle in a Sorvall™ GSA rotor, use of the Nalgene centrifuge bottle adapter (DS3125-0250) is required
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.