missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Nalgene™ Suspended Magnetic Stir Bars

Use Nalgene Floating Magnetic Stir Bars for constant low-speed stirring action in tissue culture applications.

$296.43 - $1,259.14

Specifications

Description Suspended Magnetic Stir Bar; Stirbar Floating TFE/ETFE 155 (dia.) x 31mm
View More Specs

Products 2
Catalog Number Mfr. No. For Use With (Equipment) Diameter (Metric) Height (Metric) Price Quantity  
Catalog Number Mfr. No. For Use With (Equipment) Diameter (Metric) Height (Metric) Price Quantity  
14-511-99A
View Documents
Thermo Scientific™
DS66300250
250mL beaker 54mm 28mm
Case for $296.43
 
14-511-99E
View Documents
Thermo Scientific™
DS66304000
4000mL beaker 155mm 31mm
Case of 2 Each for $1,259.14
 
Description

Description

Constantly stir tissue culture applications at a low speed with Thermo Scientific™ Nalgene™ Suspended Magnetic Stir Bars.

Features:
  • Legs can be compressed to fit through 1.5in. (38mm) openings
  • Stir bar is slightly raised off the bottom surface of the vessel to minimize grinding effect on cells
  • Consistant action at low speeds
  • Excellent resistance to chemicals and temperatures up to 150°C
  • Alnico V magnet encapsulated in PTFE with ETFE bracket

Note:

Disassemble for cleaning.

Warranty
: 90 days. For warranty terms, please contact technicalsupport@thermofisher.com

Specifications

Specifications

Suspended Magnetic Stir Bar; Stirbar Floating TFE/ETFE 155 (dia.) x 31mm
SDS

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.