missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Nalgene™ Suspended Magnetic Stir Bars
Use Nalgene Floating Magnetic Stir Bars for constant low-speed stirring action in tissue culture applications.
Manufacturer: Thermo Scientific™ DS66300250
Description
Constantly stir tissue culture applications at a low speed with Thermo Scientific™ Nalgene™ Suspended Magnetic Stir Bars.
Features:- Legs can be compressed to fit through 1.5in. (38mm) openings
- Stir bar is slightly raised off the bottom surface of the vessel to minimize grinding effect on cells
- Consistant action at low speeds
- Excellent resistance to chemicals and temperatures up to 150°C
- Alnico V magnet encapsulated in Teflon™ TFE with Tefzel™ ETFE bracket
Note:
Disassemble for cleaning.
Specifications
Suspended Magnetic Stir Bar; Stirbar Floating TFE/ETFE 54 (dia.) x 28mm | |
250mL beaker | |
28mm |
2.125 in. | |
1.125 in. | |
54mm |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.