Thermo Scientific™ Nalgene™ Sterile Analytical Filter Funnels
Easily retrieve filter membranes for growth and analysis using these funnels. They are useful for large volume microbiological QC samples.
$324.58 - $324.58
Products ${productFamilyLength}
Catalog Number Mfr. No. Pore Size Price Quantity  
Catalog Number Mfr. No. Pore Size Price Quantity  
09-740-30B Thermo Scientific™
N1404045
0.45μm Pack of 12 Each for $324.58
Case of 6 Pack for $1,947.48
Specifications
Description
Easily retrieve filter membranes for growth and analysis using the Thermo Scientific™ Nalgene™ Sterile Analytical Filter Funnels. With a convenient snap-away design, these filters are particularly useful for large volume microbiological QC samples. They are compatible with filter manifolds and filtering flasks.
Features:
  • Disposable, presterilized funnel
  • Removable 47mm diameter membrane for use with filtering flasks and manifolds
  • 0.45μm Triton™-free cellulose nitrate membrane with gray grid
  • Gamma sterilized, individually sealed in plastic bag
Includes: Upper chamber with reference mark at 150mL; polystyrene cover; CN membrane; support pad; polystyrene support plate/funnel that fits a stopper or adapter with 9/16″ hole; quick-disconnect adapter for 1/4 or 5/16″ tubing. Each pack of 12 funnels includes one vacuum gasket that replaces stopper or adapter with 9/16″ or larger opening.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.